Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metoclopramide Hydrochloride, Monohydrate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18604555 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-555 25 kg Amber Glass Bottle
18-604-550 10 g Amber Glass Bottle
18-604-551 25 g Amber Glass Bottle
18-604-552 100 g Amber Glass Bottle
18-604-553 1 kg Amber Glass Bottle
18-604-554 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-555 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ME17625KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ME176, 54143-57-6, C14H22ClN3O2·HCI·H2O

Spectrum™ Chemical Metoclopramide Hydrochloride, Monohydrate, USP is a medication used mostly for stomach and esophageal problems to treat and prevent nausea and vomiting. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54143-57-6
Molecular Formula C14H25Cl2N3O3
Molecular Weight (g/mol) 354.27
MDL Number MFCD00150634
InChI Key KJBLQGHJOCAOJP-UHFFFAOYSA-N
IUPAC Name 4-amino-5-chloro-N-[2-(diethylamino)ethyl]-2-methoxybenzamide hydrate hydrochloride
SMILES O.Cl.CCN(CC)CCNC(=O)C1=CC(Cl)=C(N)C=C1OC

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 98 to 101%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.