Learn More
Medchemexpress LLC Dapaconazole | 1269726-67-1 | 99.8% | 415.2 g/mol | C19H15Cl2F3N2O | 10 MG
SDP
Catalog No. 5000288565
Encompass_Preferred
Dapaconazole is an antifungal research compound that inhibits sterol 14α-demethylase (a cytochrome P450 enzyme). It is supplied as a solid and as a DMSO solution for in vitro studies of fungal CYP51 inhibition and antifungal activity. CAS RN 1269726-67-1; molecular formula C19H15Cl2F3N2O.
  • Inhibits sterol 14α-demethylase with an IC50 of 1.4 μM.
  • High purity (99.76%) suitable for biochemical assays.
  • Available in small solid quantities (5-100 mg) and as a 10 mM solution in DMSO for screening.
  • Commonly used in in vitro antifungal and CYP51 inhibition studies.
Catalog No. 50-002-88565 Supplier Medchemexpress LLC Supplier No. HY1671910MG
Dapaconazole is an antifungal research compound that inhibits sterol 14α-demethylase (a cytochrome P450 enzyme). It is supplied as a solid and as a DMSO solution for in vitro studies of fungal CYP51 inhibition and antifungal activity. CAS RN 1269726-67-1; molecular formula C19H15Cl2F3N2O.
  • Inhibits sterol 14α-demethylase with an IC50 of 1.4 μM.
  • High purity (99.76%) suitable for biochemical assays.
  • Available in small solid quantities (5-100 mg) and as a 10 mM solution in DMSO for screening.
  • Commonly used in in vitro antifungal and CYP51 inhibition studies.