Learn More
Medchemexpress LLC Dapaconazole | 1269726-67-1 | 99.8% | 415.24 | 5 MG
SDP
Catalog No. 5000393573
Encompass_Preferred
Dapaconazole is an antifungal agent primarily known for inhibiting sterol 14α-demethylase cytochrome P450 activity. It has shown significant bioavailability in in vivo studies.
  • Antifungal activity
  • Inhibits sterol 14α-demethylase cytochrome P450 activity
  • IC50 of 1.4 μM against CYP26
  • Demonstrates 97.3% bioavailability in male beagle dogs
Catalog No. 50-003-93573 Supplier Medchemexpress LLC Supplier No. HY167195MG
Dapaconazole is an antifungal agent primarily known for inhibiting sterol 14α-demethylase cytochrome P450 activity. It has shown significant bioavailability in in vivo studies.
  • Antifungal activity
  • Inhibits sterol 14α-demethylase cytochrome P450 activity
  • IC50 of 1.4 μM against CYP26
  • Demonstrates 97.3% bioavailability in male beagle dogs