Learn More
Medchemexpress LLC HY-10450 10mg Medchemexpress, Dapagliflozin CAS:461432-26-8 Purity:>98%
SDP
Catalog No. 502028753
Encompass_Preferred
Medchemexpress, HY-10450 10mg Dapagliflozin CAS:461432-26-8 Dapagliflozin (BMS-512148), a new type of drug used to treat diabetes mellitus (DM), is a competitive sodium/glucose cotransporter 2 (SGLT2) inhibitor, which results in excretion of glucose into the urine.Dapagliflozin induces HIF1 expression and attenuates renal IR injury[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8753 Supplier Medchemexpress LLC Supplier No. HY1045010MG
Medchemexpress, HY-10450 10mg Dapagliflozin CAS:461432-26-8 Dapagliflozin (BMS-512148), a new type of drug used to treat diabetes mellitus (DM), is a competitive sodium/glucose cotransporter 2 (SGLT2) inhibitor, which results in excretion of glucose into the urine.Dapagliflozin induces HIF1 expression and attenuates renal IR injury[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.