Learn More
Cayman Chemical Dapagliflozin, Purity: ≥98%, CAS: 461432-26-8, 10 mg
SDP
Catalog No. 502953459
Encompass_Preferred
An SGLT2 inhibitor (IC50 = 1.12 and 3 nM for the human and rat enzymes, respectively); selective for SGLT2 over SGLT1 (IC50 = 1,391 and 620 nM for the human and rat enzymes, respectively) and human adipocyte GLUT activity at 20 µM; increases urinary glucose levels in normal and Zucker diabetic rats at 0.1 and 1 mg/kg; decreases fasting and fed plasma glucose levels in Zucker diabetic rats at 0.01, 0.1, and 1 mg/kg
Catalog No. 50-295-3459 Supplier Cayman Chemical Supplier No. 1157410 MG
An SGLT2 inhibitor (IC50 = 1.12 and 3 nM for the human and rat enzymes, respectively); selective for SGLT2 over SGLT1 (IC50 = 1,391 and 620 nM for the human and rat enzymes, respectively) and human adipocyte GLUT activity at 20 µM; increases urinary glucose levels in normal and Zucker diabetic rats at 0.1 and 1 mg/kg; decreases fasting and fed plasma glucose levels in Zucker diabetic rats at 0.01, 0.1, and 1 mg/kg