Learn More
Medchemexpress LLC HY-10450 100mg , Dapagliflozin BMS-512148 CAS:461432-26-8 Purity:98%
SDP
Catalog No. 501492710
Encompass_Preferred
Medchemexpress, HY-10450 100mg Dapagliflozin BMS-512148 CAS:461432-26-8 Formula:C21H25ClO6 Purity:98% Dapagliflozin is a new pharmacological therapy for type 2 diabetes, and inhibits sodium/glucose cotransporter 2 ( SGLT2 ), which results in excretion of glucose into the urine. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2710 Supplier Medchemexpress LLC Supplier No. HY10450100MG
Medchemexpress, HY-10450 100mg Dapagliflozin BMS-512148 CAS:461432-26-8 Formula:C21H25ClO6 Purity:98% Dapagliflozin is a new pharmacological therapy for type 2 diabetes, and inhibits sodium/glucose cotransporter 2 ( SGLT2 ), which results in excretion of glucose into the urine. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.