Learn More
TARGETMOL CHEMICALS INC Dapagliflozin 2S-1 2-propanedi
SDP
Catalog No. 502878655
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dapagliflozin ((2S)-1,2-propanediol, hydrate) (BMS-512148 (2S)-1,2-propanediol, hydrate) is a selective, orally active inhibitor of the renal sodium-glucose co-transporter type 2 (SGLT2), in development for treating type 2 diabetes mellitus (T2DM). It inhibits SGLT2, responsible for at least 90% of glucose reabsorption in the kidney. Purity 99.39%
Catalog No. 50-287-8655 Supplier TARGETMOL CHEMICALS INC Supplier No. T4460500MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dapagliflozin ((2S)-1,2-propanediol, hydrate) (BMS-512148 (2S)-1,2-propanediol, hydrate) is a selective, orally active inhibitor of the renal sodium-glucose co-transporter type 2 (SGLT2), in development for treating type 2 diabetes mellitus (T2DM). It inhibits SGLT2, responsible for at least 90% of glucose reabsorption in the kidney. Purity 99.39%