Learn More
Apexbio Technology LLC Dapagliflozin 2S Propanedio
SDP
Catalog No. 5000897549
Encompass_Preferred
Dapagliflozin ((2S)-1 2-propanediol hydrate) (CAS No 960404-48-2) is an antidiabetic agent categorized under SGLT2 inhibitors primarily researched for its role in glucose regulation Originating from studies to manage hyperglycemia in type 2 diabetes it works by inhibiting the sodium-glucose cotransporter 2 (SGLT2) in the proximal renal tubules effectively reducing reabsorption of glucose and promoting its excretion via urine Its in vitro activity is characterized by potent inhibitory effects in the nanomolar range Dapagliflozin is frequently used in cellular models and animal studies to explore glucose metabolism and evaluate glycemic control strategies Additionally it is utilized in drug screening for diabetes treatment research Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97549 Supplier Apexbio Technology LLC Supplier No. C63675
Dapagliflozin ((2S)-1 2-propanediol hydrate) (CAS No 960404-48-2) is an antidiabetic agent categorized under SGLT2 inhibitors primarily researched for its role in glucose regulation Originating from studies to manage hyperglycemia in type 2 diabetes it works by inhibiting the sodium-glucose cotransporter 2 (SGLT2) in the proximal renal tubules effectively reducing reabsorption of glucose and promoting its excretion via urine Its in vitro activity is characterized by potent inhibitory effects in the nanomolar range Dapagliflozin is frequently used in cellular models and animal studies to explore glucose metabolism and evaluate glycemic control strategies Additionally it is utilized in drug screening for diabetes treatment research Experimental concentrations typically depend on the specific research design