Learn More
Selleck Chemical LLC Dapagliflozin 5mg 461432-26-8 BMS-512148
SDP
Catalog No. 501361377
Encompass_Preferred
Dapagliflozin (BMS-512148) is a potent and selective hSGLT2 inhibitor with EC50 of 1.1 nM, exhibiting 1200-fold selectivity over hSGLT1. Phase 4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1377 Supplier Selleck Chemical LLC Supplier No. S15485MG
Dapagliflozin (BMS-512148) is a potent and selective hSGLT2 inhibitor with EC50 of 1.1 nM, exhibiting 1200-fold selectivity over hSGLT1. Phase 4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.