Learn More
Medchemexpress LLC Dapagliflozin-d5 | 1204219-80-6 | MFCD21363576 | 98.1% | 413.90 | C21H20D5ClO6 | 5 MG

Supplier: Medchemexpress LLC HY10450S5MG
Dapagliflozin-d5 is a deuterium-labeled analogue of dapagliflozin intended for use as an isotope-labeled internal standard and tracer in analytical and pharmacokinetic studies. It is supplied as a powder for preparation of calibration standards and supports accurate quantification of parent compound in biological and synthetic matrices.
- Deuterium-labeled internal standard for analytical quantification.
- Suitable for GC- and LC-MS method development and validation.
- High reported purity (98.06%).
- Molecular weight 413.90 and formula C21H20D5ClO6.
- Soluble in DMSO, DMF, and ethanol at ≥30 mg/mL.
- Powder form with recommended cold storage for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.