Learn More
Medchemexpress LLC Dapagliflozin-d5 | 1204219-80-6 | MFCD21363576 | 98.1% | 413.90 g/mol | C21H20D5ClO6 | 1 MG
SDP
Catalog No. 5000244319
Encompass_Preferred
Dapagliflozin-d5 is the pentadeuterated isotopologue of dapagliflozin used as an internal standard or tracer in quantitative mass spectrometry and analytical assays. The deuterium labeling produces a defined mass shift that enables accurate, matrix-matched quantification in bioanalytical and pharmacokinetic studies.
  • Deuterium labeled d5 internal standard for LC-MS and GC-MS.
  • High purity (98.1%) suitable for quantitative analysis.
  • Defined molecular weight for reliable mass-shift monitoring.
  • Provided in a small, research-grade package for analytical workflows.
  • Useful as a tracer in pharmacokinetic and bioanalytical studies.
Catalog No. 50-002-44319 Supplier Medchemexpress LLC Supplier No. HY10450S1MG
Dapagliflozin-d5 is the pentadeuterated isotopologue of dapagliflozin used as an internal standard or tracer in quantitative mass spectrometry and analytical assays. The deuterium labeling produces a defined mass shift that enables accurate, matrix-matched quantification in bioanalytical and pharmacokinetic studies.
  • Deuterium labeled d5 internal standard for LC-MS and GC-MS.
  • High purity (98.1%) suitable for quantitative analysis.
  • Defined molecular weight for reliable mass-shift monitoring.
  • Provided in a small, research-grade package for analytical workflows.
  • Useful as a tracer in pharmacokinetic and bioanalytical studies.