Learn More
Medchemexpress LLC HY-128723 5mg Medchemexpress, Dapagliflozin impurity CAS:960404-86-8 Purity:>98%
SDP
Catalog No. 501969765
Encompass_Preferred
Medchemexpress, HY-128723 5mg Dapagliflozin impurity CAS:960404-86-8 Dapagliflozin impurity is an enantiomer of Dapagliflozin which is a sodium-glucose transporter 2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9765 Supplier Medchemexpress LLC Supplier No. HY1287235MG
Medchemexpress, HY-128723 5mg Dapagliflozin impurity CAS:960404-86-8 Dapagliflozin impurity is an enantiomer of Dapagliflozin which is a sodium-glucose transporter 2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.