Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dapagliflozin impurity | 960404-86-8 | 99.6% | 10 MM 1 ML
SDP

Catalog No. 5000339402
Encompass_Preferred

Dapagliflozin impurity is an enantiomer of Dapagliflozin, functioning as a sodium-glucose transporter 2 inhibitor. This compound has a molecular weight of 424.87 and a chemical formula of C21H25ClO7. It is supplied as a 10 mM solution in 1 mL of DMSO, with a purity of 99.6%. It appears as an off-white to light yellow solid and is intended for laboratory research use only.

  • Enantiomer of dapagliflozin
  • Functions as a sodium-glucose transporter 2 inhibitor
  • Intended for research use only
  • Molecular weight of 424.87
  • Chemical formula: C21H25ClO7
  • Appearance: off-white to light yellow solid
  • Purity of 99.6%
  • Supplied as 10 mM in 1 mL of DMSO
  • Soluble in DMSO at 25 mg/mL (58.84 mM)
  • Recommended storage conditions:
    • Powder: -20°C for 3 years; 4°C for 2 years
    • In solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-003-39402 Supplier Medchemexpress LLC Supplier No. HY12872310MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dapagliflozin impurity is an enantiomer of Dapagliflozin, functioning as a sodium-glucose transporter 2 inhibitor. This compound has a molecular weight of 424.87 and a chemical formula of C21H25ClO7. It is supplied as a 10 mM solution in 1 mL of DMSO, with a purity of 99.6%. It appears as an off-white to light yellow solid and is intended for laboratory research use only.

  • Enantiomer of dapagliflozin
  • Functions as a sodium-glucose transporter 2 inhibitor
  • Intended for research use only
  • Molecular weight of 424.87
  • Chemical formula: C21H25ClO7
  • Appearance: off-white to light yellow solid
  • Purity of 99.6%
  • Supplied as 10 mM in 1 mL of DMSO
  • Soluble in DMSO at 25 mg/mL (58.84 mM)
  • Recommended storage conditions:
    • Powder: -20°C for 3 years; 4°C for 2 years
    • In solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.