Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Dapagliflozin

Catalog No. AC464620010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464620010 1 g
AC464620050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC464620010 Supplier Thermo Scientific Chemicals Supplier No. 464620010
Add to Cart

CAS: 461432-26-8 | C21H25ClO6 | 408.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461432-26-8
Molecular Formula C21H25ClO6
Molecular Weight (g/mol) 408.88
InChI Key JVHXJTBJCFBINQ-UHFFFAOYNA-N
IUPAC Name 2-{4-chloro-3-[(4-ethoxyphenyl)methyl]phenyl}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCOC1=CC=C(CC2=C(Cl)C=CC(=C2)C2OC(CO)C(O)C(O)C2O)C=C1

Specifications

Melting Point 65.0°C to 70.0°C
Quantity 1 g
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol
Formula Weight 408.88
Chemical Name or Material Dapagliflozin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement Call a POISON CENTER or doctor/physician if you feel unwell.

IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.