Learn More
Cayman Chemical Daphn genkwa GenkwnIn 100mg
SDP
Catalog No. 502913681
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits proliferation of MDA-MB-231 and MCF-7, Eca-109, and A549 cells (IC50s = 58.54, 101.4, 98.2, and 120.77 µM, respectively); reverses MPP+-induced increases in apoptosis, LDH release, and ROS production in an in vitro model of Parkinson’s disease inflammation using SH-SY5Y cells at 20-40 µM
Catalog No. 50-291-3681 Supplier Cayman Chemical Supplier No. 35229100 MG
A flavonoid with diverse biological activities; inhibits proliferation of MDA-MB-231 and MCF-7, Eca-109, and A549 cells (IC50s = 58.54, 101.4, 98.2, and 120.77 µM, respectively); reverses MPP+-induced increases in apoptosis, LDH release, and ROS production in an in vitro model of Parkinson’s disease inflammation using SH-SY5Y cells at 20-40 µM