Learn More
Medchemexpress LLC HY-N0281 10mg Medchemexpress, Daphnetin CAS:486-35-1 Purity:>98%
SDP
Catalog No. 501933505
Encompass_Preferred
Medchemexpress, HY-N0281 10mg Daphnetin CAS:486-35-1 Daphnetin (7,8-dihydroxycoumarin), one coumarin derivative isolated from plants of the Genus Daphne, is a protein kinase inhibitor, with IC 50 s of 7.67 μM, 9.33 μM and 25.01 μM for EGFR, PKA and PKC in vitro, respectively [1] [2] . Daphnetin (7,8-dihydroxycoumarin) is a secondary metabolite of plants used in folk medicine to counter inflammatory and allergic diseases, also has been clinically used in the treatment of coagulation disorders, rheumatoid arthritis with anti-malarian and anti-pyretic properties [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3505 Supplier Medchemexpress LLC Supplier No. HYN028110MG
Medchemexpress, HY-N0281 10mg Daphnetin CAS:486-35-1 Daphnetin (7,8-dihydroxycoumarin), one coumarin derivative isolated from plants of the Genus Daphne, is a protein kinase inhibitor, with IC 50 s of 7.67 μM, 9.33 μM and 25.01 μM for EGFR, PKA and PKC in vitro, respectively [1] [2] . Daphnetin (7,8-dihydroxycoumarin) is a secondary metabolite of plants used in folk medicine to counter inflammatory and allergic diseases, also has been clinically used in the treatment of coagulation disorders, rheumatoid arthritis with anti-malarian and anti-pyretic properties [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.