Learn More
Apexbio Technology LLC DAPHNETIN-20MG
SDP
Catalog No. 5000567778
Encompass_Preferred
Daphnetin (CAS No 486-35-1) is a naturally occurring coumarin derivative primarily used in biomedical research It is derived from different plants including Daphne and Wikstroemia species The compound is known to inhibit various inflammatory pathways and has been shown to modulate enzymes like protein kinases Daphnetin exhibits significant in vitro activity in the nanomolar to low micromolar range It is commonly applied in cancer research studying its effects on tumor cell proliferation and apoptosis and in neurological studies focusing on neuroprotection Typical research applications include cell models animal studies and drug screening assays Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67778 Supplier Apexbio Technology LLC Supplier No. N156720
Daphnetin (CAS No 486-35-1) is a naturally occurring coumarin derivative primarily used in biomedical research It is derived from different plants including Daphne and Wikstroemia species The compound is known to inhibit various inflammatory pathways and has been shown to modulate enzymes like protein kinases Daphnetin exhibits significant in vitro activity in the nanomolar to low micromolar range It is commonly applied in cancer research studying its effects on tumor cell proliferation and apoptosis and in neurological studies focusing on neuroprotection Typical research applications include cell models animal studies and drug screening assays Experimental concentrations typically depend on the specific research design