Learn More
Enzo Life Sciences Daphnetin (25mg). CAS: 486-35-1
SDP
Supplier:  Enzo Life Sciences ALX270281M025
Encompass_Preferred
Inhibitor of protein kinases. EGF receptor tyrosine kinase (IC50=7.87µM), PKA (IC50=9.33µM), PKC (IC50=25.01µM). Antioxidant. Antimalarial agent. Alternative name: 7,8-Dihydroxycoumarin. Formula: C9H6O4. MW: 178.1. Purity: ≥97% (HPLC). Solubility: Soluble in 100% ethanol, methanol or dimethyl formamide. Long Term Storage: Ambient.
Catalog No. 50-200-8459