Learn More
TARGETMOL CHEMICALS INC Daphnetin 50MG
SDP
Catalog No. 502876531
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Daphnetin (7,8-Dihydroxycoumarin), a natural coumarin derivative, is a protein kinase inhibitor with inhibitory for EGFR (IC50 7.67 uM), PKA (IC50 9.33 uM), and PKC (IC50 25.01 uM), also exhibit anti-oxidant and anti-inflammatory activities. Purity 99.82%
Catalog No. 50-287-6531 Supplier TARGETMOL CHEMICALS INC Supplier No. T285150MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Daphnetin (7,8-Dihydroxycoumarin), a natural coumarin derivative, is a protein kinase inhibitor with inhibitory for EGFR (IC50 7.67 uM), PKA (IC50 9.33 uM), and PKC (IC50 25.01 uM), also exhibit anti-oxidant and anti-inflammatory activities. Purity 99.82%