Learn More
Selleck Chemical LLC Daphnetin 10mg 486-35-1
SDP
Supplier:  Selleck Chemical LLC S255410MG
Encompass_Preferred
Daphnetin, a natural coumarin derivative, is a protein kinase inhibitor, inhibits EGFR, PKA and PKC with IC50 of 7.67 ?M, 9.33 ?M and 25.01 ?M, respectively, also known to exhibit anti-inflammatory and anti-oxidant activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-796-0