Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Daphnin | 486-55-5 | 5 MG
SDP

Catalog No. 5000417252
Encompass_Preferred

Daphnin is one of the major coumarin bioactive components with antibacterial activity. It is isolated from the whole herb of Daphne odora (Thunb.), a folk medicine in China used for fever relief.

  • Antibacterial activity
  • Isolated from Daphne odora (Thunb.)
  • Used in folk medicine for fever relief
  • Purity: 99.85%
  • Molecular weight: 340.28
  • Formula: C15H16O9
  • Appearance: Solid, light brown to khaki
  • Target: Bacterial
  • Pathway: Anti-infection
  • Melting/freezing point: 216-217°C

Catalog No. 50-004-17252 Supplier Medchemexpress LLC Supplier No. HYN72525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Daphnin is one of the major coumarin bioactive components with antibacterial activity. It is isolated from the whole herb of Daphne odora (Thunb.), a folk medicine in China used for fever relief.

  • Antibacterial activity
  • Isolated from Daphne odora (Thunb.)
  • Used in folk medicine for fever relief
  • Purity: 99.85%
  • Molecular weight: 340.28
  • Formula: C15H16O9
  • Appearance: Solid, light brown to khaki
  • Target: Bacterial
  • Pathway: Anti-infection
  • Melting/freezing point: 216-217°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.