Learn More
Medchemexpress LLC HY-N0699 5mg Medchemexpress, Daphnoretin CAS:2034-69-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06995MG
Encompass_Preferred
Medchemexpress, HY-N0699 5mg Daphnoretin CAS:2034-69-7 Daphnoretin (Dephnoretin), isolated from Wikstroemia indica, possesses antiviral activity. Daphnoretin likes PMA, may direct activation of protein kinase C which in turn activated NADPH oxidase and elicited respiratory burst. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0304