Learn More
Supply Solutions DAPI, 28718-90-3, MFCD00012681, 10mg
Catalog No. 501003350
Encompass
Empirical Formula (Hill Notation) C16H15N5 2HCl, Molecular Weight 350.25, Synonym: 2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, 4?,6-Diamidino-2-phenylindole dihydrochloride, DAPI dihydrochloride; DAPI is a fluorescent dye that binds selectively to double-stranded DNA and forms strongly fluorescent DNA-DAPI complexes with high specificity.
Catalog No. 50-100-3350 Supplier Supply Solutions Supplier No. SIGMA 10236276001
Empirical Formula (Hill Notation) C16H15N5 2HCl, Molecular Weight 350.25, Synonym: 2-(4-Amidinophenyl)-6-indolecarbamidine dihydrochloride, 4?,6-Diamidino-2-phenylindole dihydrochloride, DAPI dihydrochloride; DAPI is a fluorescent dye that binds selectively to double-stranded DNA and forms strongly fluorescent DNA-DAPI complexes with high specificity.