Learn More
AnaSpec DAPI[[4'',6-Diamidino-2-phenylindole, dihydrochloride]] | MW 350.3 | (10 mg)
Catalog No. 50850585
Encompass
DAPI[[4'',6-Diamidino-2-phenylindole, dihydrochloride]] ; MW 350.3; (10 mg) AT-selective minor groove binder that exhibits nice fluorescence enhancement in the presence of DNA.
Catalog No. 50-850-585 Supplier AnaSpec Supplier No. AS83210
DAPI[[4'',6-Diamidino-2-phenylindole, dihydrochloride]] ; MW 350.3; (10 mg) AT-selective minor groove binder that exhibits nice fluorescence enhancement in the presence of DNA.