Learn More
AdipoGen DAPI 2HCl Solution 10mM water
SDP
Catalog No. 502561011
Encompass_Preferred
Chemical. CAS 28718-90-3. Formula C16H15N5 . 2HCl. MW 350.25. DAPI 4,6-Diamidino-2-phenylindole dihydrochloride is a cell permeable, fluorescent dye that is a minor groove-binding probe for DNA. Binds to the minor groove of double-stranded DNA preferentially to AT rich DNA, forming a stable complex which fluoresces approximately 20 times greater than DAPI alone. DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels. It may be used for photofootprinting of DNA, to detect annealed probes in blotting applications by specifically visualizing the double-stranded complex, and to study the changes in DNA and analyze DNA content during apoptosis using flow cytometry. DAPI staining has also been shown to be a sensitive and specific detection method for mycoplasma. Spectral Data Excitation lambdaex 340 nm Emission lambdaem 488 nm only DAPI. Excitation lambdaex 360nm Emission lambdaem 460nm DAPI-DNA complex.
Catalog No. 50-256-1011 Supplier AdipoGen Supplier No. CDXD1217M005
Chemical. CAS 28718-90-3. Formula C16H15N5 . 2HCl. MW 350.25. DAPI 4,6-Diamidino-2-phenylindole dihydrochloride is a cell permeable, fluorescent dye that is a minor groove-binding probe for DNA. Binds to the minor groove of double-stranded DNA preferentially to AT rich DNA, forming a stable complex which fluoresces approximately 20 times greater than DAPI alone. DAPI is several times more sensitive than ethidium bromide for staining DNA in agarose gels. It may be used for photofootprinting of DNA, to detect annealed probes in blotting applications by specifically visualizing the double-stranded complex, and to study the changes in DNA and analyze DNA content during apoptosis using flow cytometry. DAPI staining has also been shown to be a sensitive and specific detection method for mycoplasma. Spectral Data Excitation lambdaex 340 nm Emission lambdaem 488 nm only DAPI. Excitation lambdaex 360nm Emission lambdaem 460nm DAPI-DNA complex.