Learn More
Diagnostic Biosystems DAPI
Catalog No. 502407445
Encompass_Preferred
DAPI or 4′,6-diamidino-2-phenylindole, is a fluorescent stain that binds strongly to adenine–thymine-rich regions in DNA. It is used extensively in fluorescence microscopy. As DAPI can pass through an intact cell membrane, it can be used to stain both live and fixed cells.\n\nFeatures\n• This DAPI solution is optimized for use on formalin-fixed, paraffin-embedded tissue sections.\n• The DAPI solution contains an anti-freeze agent so the reagent can be stored at -20 °C without freezing.\n• For convenience the DAPI solution can be removed from the freezer and used immediately without the necessity of thawing.\n• There is no need to aliquot and freeze small portions.\n\nKnown Applications\n• Fluorescence In Situ Hybridization\n• Immunofluorescence with fluorescently labeled antibodies or probes.\n• Fluorescent staining methods.
Catalog No. 50-240-7445 Supplier Diagnostic Biosystems Supplier No. K108
DAPI or 4′,6-diamidino-2-phenylindole, is a fluorescent stain that binds strongly to adenine–thymine-rich regions in DNA. It is used extensively in fluorescence microscopy. As DAPI can pass through an intact cell membrane, it can be used to stain both live and fixed cells.\n\nFeatures\n• This DAPI solution is optimized for use on formalin-fixed, paraffin-embedded tissue sections.\n• The DAPI solution contains an anti-freeze agent so the reagent can be stored at -20 °C without freezing.\n• For convenience the DAPI solution can be removed from the freezer and used immediately without the necessity of thawing.\n• There is no need to aliquot and freeze small portions.\n\nKnown Applications\n• Fluorescence In Situ Hybridization\n• Immunofluorescence with fluorescently labeled antibodies or probes.\n• Fluorescent staining methods.