Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ DAPI

Catalog No. 89941409 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-941-409 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-941-409 Supplier Abnova™ Supplier No. U0331
Add to Cart
Add to Cart

DAPI is a nuclear and chromosome counterstain, it emits blue fluorescence upon binding to AT regions of DNA. Although the dye is cell impermeant, higher concentrations will enter a live cell.

Specifications

Content And Storage Store at room temperature and protect from light.
For Use With (Application) Cell Staining
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.