Learn More
Medchemexpress LLC HY-D0814 10mg Medchemexpress, DAPI (dihydrochloride) CAS:28718-90-3 Purity:>98%
SDP
Catalog No. 502029411
Encompass_Preferred
Medchemexpress, HY-D0814 10mg DAPI (dihydrochloride) CAS:28718-90-3 DAPI dihydrochloride (4',6-diamidino-2-phenylindole) is a fluorescent stain by binding in the minor grove of A-T rich sequences of DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9411 Supplier Medchemexpress LLC Supplier No. HYD081410MG
Medchemexpress, HY-D0814 10mg DAPI (dihydrochloride) CAS:28718-90-3 DAPI dihydrochloride (4',6-diamidino-2-phenylindole) is a fluorescent stain by binding in the minor grove of A-T rich sequences of DNA. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.