Learn More
TARGETMOL CHEMICALS INC DAPI DIHYDROCHLORIDE 25MG
SDP
Catalog No. 502577255
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DAPI Dihydrochloride (FxCycle Violet) is a cell-permeable fluorescent probe by binding in the minor grove of A-T rich sequences of DNA used to stain DNA and chromosomes with a preference for adenine and thymine rich DNA. purity: 99%
Catalog No. 50-257-7255 Supplier TARGETMOL CHEMICALS INC Supplier No. T1982725MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DAPI Dihydrochloride (FxCycle Violet) is a cell-permeable fluorescent probe by binding in the minor grove of A-T rich sequences of DNA used to stain DNA and chromosomes with a preference for adenine and thymine rich DNA. purity: 99%