Learn More
Medchemexpress LLC DAPI dihydrochloride (4',6-diamidino-2-phenylindole dihydrochloride) | 28718-90-3 | 99.9% | 350.25 | 25 ML
SDP
Catalog No. 5000429609
Encompass_Preferred
AntiFade Reagent (with DAPI) is designed to mitigate fluorescence quenching, effectively preserving the intensity of various common fluorescent dyes. It offers a simple operational procedure and delivers a strong anti-fluorescence quenching effect. This reagent also incorporates DAPI, making it suitable for staining cell nuclei.
  • Slows fluorescence quenching.
  • Simple operation.
  • Good anti-fluorescence quenching effect.
  • Contains DAPI for cell nucleus staining.
  • Suitable for nuclear localization studies.
Catalog No. 50-004-29609 Supplier Medchemexpress LLC Supplier No. 25MLHYK1048
AntiFade Reagent (with DAPI) is designed to mitigate fluorescence quenching, effectively preserving the intensity of various common fluorescent dyes. It offers a simple operational procedure and delivers a strong anti-fluorescence quenching effect. This reagent also incorporates DAPI, making it suitable for staining cell nuclei.
  • Slows fluorescence quenching.
  • Simple operation.
  • Good anti-fluorescence quenching effect.
  • Contains DAPI for cell nucleus staining.
  • Suitable for nuclear localization studies.