Learn More
Selleck Chemical LLC DAPI Dihydrochloride-5mg
SDP
Supplier:  Selleck Chemical LLC S99805MG
Encompass_Preferred
DAPI Dihydrochloride (DAPI 2HCl) is a fluorescent dye that binds strongly to adenine–thymine-rich regions in DNA.
Catalog No. 50-239-0065