Learn More
DAPI, Dilactate, Sigma-Aldrich
Catalog No. 01258838 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
01-258-838 10 mg
1 options
Catalog No. 01-258-838 Supplier MilliporeSigma Supelco Supplier No. D956410MG
Add to Cart
Add to Cart
A high sensitivity dye used to detect single nucleic acid molecules. DAPI has been shown to detect a single DNA molecule in solution, and to detect femtograms of DNA in single cells and chloroplasts.
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless to Yellow
Format Bottle
Quantity 10 mg
Product Type Stain