Learn More
Electron Microscopy Sciences Dapi Fluoromount G 20ML
SDP
Catalog No. 5034716
Encompass_Preferred
DAPI-Fluoromont G is a water-soluble, instant blue nuclear probe fluorescing (455nm) compound for mounting fixed slides using a staining procedure having an aqueous final step. Mounting slides with DAPI Fluoromont G stains the cell nucleus and may reduce fluorochrome quenching during analysis of slides by fluorescence microscopy. this mounting medium also provides a semi permanent seal for long term storage of slide preparations.
Catalog No. 50-347-16 Supplier Electron Microscopy Sciences Supplier No. 1798424
DAPI-Fluoromont G is a water-soluble, instant blue nuclear probe fluorescing (455nm) compound for mounting fixed slides using a staining procedure having an aqueous final step. Mounting slides with DAPI Fluoromont G stains the cell nucleus and may reduce fluorochrome quenching during analysis of slides by fluorescence microscopy. this mounting medium also provides a semi permanent seal for long term storage of slide preparations.