Learn More
Enzo Life Sciences DAPI (ultra pure) (100mg). CAS: 28718-90-3
SDP
Catalog No. 502012639
Encompass_Preferred
DAPI is a fluorescent dye that binds nucleic acids and demonstrates a high affinity for DNA. It is used extensively in fluorescence microscopy, flow cytometry and microplate assays. DAPI is cell permeable and is readily used for detecting both live, dead and fixed cells. DAPI emits in the blue spectral range and is ideal for multiplexed assays inclusive of green-fluorescent molecules like fluorescein and green fluorescent protein (GFP), or red-fluorescent stains like Texas Red. Aside from labeling cell nuclei, DAPI is also used for the detection of mycoplasma or viral DNA in cell cultures. Wavelength Maxima: Excitation 358nm, Emission 461nm Alternative name: 4',6-Diamidino-2-phenylindole . dihydrochloride. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-2639 Supplier Enzo Life Sciences Supplier No. ENZ52404
DAPI is a fluorescent dye that binds nucleic acids and demonstrates a high affinity for DNA. It is used extensively in fluorescence microscopy, flow cytometry and microplate assays. DAPI is cell permeable and is readily used for detecting both live, dead and fixed cells. DAPI emits in the blue spectral range and is ideal for multiplexed assays inclusive of green-fluorescent molecules like fluorescein and green fluorescent protein (GFP), or red-fluorescent stains like Texas Red. Aside from labeling cell nuclei, DAPI is also used for the detection of mycoplasma or viral DNA in cell cultures. Wavelength Maxima: Excitation 358nm, Emission 461nm Alternative name: 4',6-Diamidino-2-phenylindole . dihydrochloride. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO. Long Term Storage: -20°C.