Learn More
Cayman Chemical DAPI (hydrochloride), CAS: 28718-90-3, 10 mg
SDP
Catalog No. 502955317
Encompass_Preferred
A fluorescent probe which is commonly used to stain DNA and chromosomes for fluorescent microscopy and flow cytometry applications; characterized by ultraviolet (max 358 nm) excitation and blue (max 461 nm) emission wavelengths
Catalog No. 50-295-5317 Supplier Cayman Chemical Supplier No. 1428510 MG
A fluorescent probe which is commonly used to stain DNA and chromosomes for fluorescent microscopy and flow cytometry applications; characterized by ultraviolet (max 358 nm) excitation and blue (max 461 nm) emission wavelengths