Learn More
Selleck Chemical LLC Dapiprazole Hydrochloride S5281-25mg
SDP
Catalog No. 503112025
Encompass_Preferred
Dapiprazole Hydrochloride (Glamidolo Hydrochloride Reversil Hydrochloride) is the hydrochloride salt form of dapiprazole an alpha-adrenergic blocker used to reverse mydriasis after eye examination
Catalog No. 50-311-2025 Supplier Selleck Chemical LLC Supplier No. S528125MG
Dapiprazole Hydrochloride (Glamidolo Hydrochloride Reversil Hydrochloride) is the hydrochloride salt form of dapiprazole an alpha-adrenergic blocker used to reverse mydriasis after eye examination