Learn More
Medchemexpress LLC Dapiprazole hydrochloride | 72822-13-0 | 99.3% | 361.91 | C19H28ClN5 | 10 MG
SDP
Catalog No. 5000298690
Encompass_Preferred
Dapiprazole hydrochloride is a potent, selective alpha-1 adrenoceptor antagonist supplied as a solid for laboratory research. It is used experimentally to reverse mydriasis, to study adrenergic signaling, and in preclinical investigations of opioid withdrawal. Handle according to the product SDS and institutional safety protocols.
  • High purity: 99.3% by assay.
  • Solid, white to off-white form suitable for accurate weighing.
  • Available in small milligram quantities for research workflows.
  • Well characterized: CAS 72822-13-0; molecular weight 361.91.
  • Intended for laboratory research use; consult SDS for handling and storage.
Catalog No. 50-002-98690 Supplier Medchemexpress LLC Supplier No. HYA0142A10MG
Dapiprazole hydrochloride is a potent, selective alpha-1 adrenoceptor antagonist supplied as a solid for laboratory research. It is used experimentally to reverse mydriasis, to study adrenergic signaling, and in preclinical investigations of opioid withdrawal. Handle according to the product SDS and institutional safety protocols.
  • High purity: 99.3% by assay.
  • Solid, white to off-white form suitable for accurate weighing.
  • Available in small milligram quantities for research workflows.
  • Well characterized: CAS 72822-13-0; molecular weight 361.91.
  • Intended for laboratory research use; consult SDS for handling and storage.