Learn More
Medchemexpress LLC Dapiprazole hydrochloride | 72822-13-0 | 99.3% | C19H28ClN5 | 25 MG
SDP
Catalog No. 5000359084
Encompass_Preferred
Dapiprazole hydrochloride is a potent, selective, and orally active alpha-1 adrenoceptor antagonist. It suppresses opioid withdrawal symptoms and is also utilized in eye drops to reverse mydriasis.
  • Potent, selective, and orally active alpha-1 adrenoceptor antagonist
  • Suppresses opioid withdrawal symptoms
  • Used in eye drops for reversing mydriasis
Catalog No. 50-003-59084 Supplier Medchemexpress LLC Supplier No. 25MGHYA0142A
Dapiprazole hydrochloride is a potent, selective, and orally active alpha-1 adrenoceptor antagonist. It suppresses opioid withdrawal symptoms and is also utilized in eye drops to reverse mydriasis.
  • Potent, selective, and orally active alpha-1 adrenoceptor antagonist
  • Suppresses opioid withdrawal symptoms
  • Used in eye drops for reversing mydriasis