Learn More
Medchemexpress LLC Dapiprazole hydrochloride | 72822-13-0 | 99.3% | C19H28ClN5 | 50 MG
SDP
Catalog No. 5000432812
Encompass_Preferred
Dapiprazole hydrochloride is a potent, selective, and orally active alpha-1 adrenoceptor antagonist.
  • Suppresses opioid withdrawal symptoms.
  • Used as eye drops for reversing mydriasis.
Catalog No. 50-004-32812 Supplier Medchemexpress LLC Supplier No. HYA0142A50MG
Dapiprazole hydrochloride is a potent, selective, and orally active alpha-1 adrenoceptor antagonist.
  • Suppresses opioid withdrawal symptoms.
  • Used as eye drops for reversing mydriasis.