Learn More
Selleck Chemical LLC Dapivirine (TMC120) S2914-5mg
SDP
Catalog No. 503108169
Encompass_Preferred
Dapivirine (TMC120) is a non-nucleoside inhibitor for HIV reverse transcriptase with IC50 of 24 nM inhibits a broad panel of HIV-1 isolates from different classes inclucing a wide range of NNRTI-resistant isolates Phase 3
Catalog No. 50-310-8169 Supplier Selleck Chemical LLC Supplier No. S29145MG
Dapivirine (TMC120) is a non-nucleoside inhibitor for HIV reverse transcriptase with IC50 of 24 nM inhibits a broad panel of HIV-1 isolates from different classes inclucing a wide range of NNRTI-resistant isolates Phase 3