Learn More
Carnabio Usa Inc DAPK1/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 021345
Encompass
DAPK1, Catalytic domain, Wild type, Amino Acid 1-289, NP_004929.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0390