Learn More
Eurofins Discoverx DAPK1, active, 10 µg. (14-692)
Supplier:  Eurofins Discoverx 14692
Encompass
N-terminal GST-tagged recombinant, human DAPK1 amino acids 1-296. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9199