Learn More
Aviva Systems Biology Corp DAPK1 Antibody - N-terminal region (ARP58219_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58219_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAPK1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DAPK1. Alternate names: DAPK, DKFZp781I035ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62262