Learn More
Selleck Chemical LLC Daporinad (FK866) S2799-50mg
SDP
Catalog No. 503112630
Encompass_Preferred
Daporinad (FK866) effectively inhibits nicotinamide phosphoribosyltransferase (NMPRTase Nampt) with IC50 of 0 09 nM in a cell-free assay Daporinad (FK866 APO866) triggers autophagy Phase 1/2
Catalog No. 50-311-2630 Supplier Selleck Chemical LLC Supplier No. S279950MG
Daporinad (FK866) effectively inhibits nicotinamide phosphoribosyltransferase (NMPRTase Nampt) with IC50 of 0 09 nM in a cell-free assay Daporinad (FK866 APO866) triggers autophagy Phase 1/2