Learn More
Medchemexpress LLC Dapoxetine-d7 hydrochloride | 2699607-47-9 | 100.0% | 1 MG
SDP
Catalog No. 5000406785
Encompass_Preferred
Dapoxetine-d7 hydrochloride is the deuterium labeled form of Dapoxetine hydrochloride, a short-acting selective serotonin reuptake inhibitor (SSRI). This compound is intended for research use only.
  • Utilized as a tracer
  • Serves as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
  • Short-acting selective serotonin reuptake inhibitor (SSRI)
Catalog No. 50-004-06785 Supplier Medchemexpress LLC Supplier No. HYB0304AS1MG
Dapoxetine-d7 hydrochloride is the deuterium labeled form of Dapoxetine hydrochloride, a short-acting selective serotonin reuptake inhibitor (SSRI). This compound is intended for research use only.
  • Utilized as a tracer
  • Serves as an internal standard for quantitative analysis through techniques such as NMR, GC-MS, or LC-MS
  • Short-acting selective serotonin reuptake inhibitor (SSRI)