Learn More
Apexbio Technology LLC Dapoxetine HCl 129938-20-1 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092126
Encompass_Preferred
Dapoxetine hydrochloride (CAS 129938-20-1) is a short-acting selective serotonin reuptake inhibitor (SSRI) It exerts its pharmacological effect by inhibiting the reuptake of serotonin (5-HT) at the presynaptic membrane thereby increasing serotonin activity in the synaptic cleft This modulation of serotonergic neurotransmission has been shown to delay ejaculation latency making dapoxetine hydrochloride an established model compound for investigating serotonergic regulation of ejaculatory control It is widely used in research on the neurobiology and therapeutic approaches to premature ejaculation and related disorders
Catalog No. 50-000-92126 Supplier Apexbio Technology LLC Supplier No. B22645.1
Dapoxetine hydrochloride (CAS 129938-20-1) is a short-acting selective serotonin reuptake inhibitor (SSRI) It exerts its pharmacological effect by inhibiting the reuptake of serotonin (5-HT) at the presynaptic membrane thereby increasing serotonin activity in the synaptic cleft This modulation of serotonergic neurotransmission has been shown to delay ejaculation latency making dapoxetine hydrochloride an established model compound for investigating serotonergic regulation of ejaculatory control It is widely used in research on the neurobiology and therapeutic approaches to premature ejaculation and related disorders