Learn More
Medchemexpress LLC Dapoxetine hydrochloride | 129938-20-1 | 100.0% | 1 ML
Supplier: Medchemexpress LLC HYB0304A10MM/1ML
Dapoxetine (LY-210448) hydrochloride is an orally active and selective serotonin reuptake inhibitor (SSRI) that can be utilized for research related to premature ejaculation (PE).
- Orally active and selective serotonin reuptake inhibitor (SSRI).
- Applicable for research of premature ejaculation (PE).
- Binds to 5-HT, norepinephrine, and dopamine reuptake transporters.
- Inhibits 5-HT, norepinephrine, and dopamine uptake with an order of potency: 5-HT > norepinephrine ≈ dopamine.
- Demonstrates an IC50 of 1.12 nM for inhibiting the uptake of [3H]5-HT by the 5-HT reuptake transporter.
- Shows IC50 values of 202 nM and 1720 nM for inhibiting the uptake of [3H]norepinephrine and [3H]dopamine, respectively.
- Significantly inhibits testosterone-mediated increase in prostate weight and attenuates testosterone-induced prostatic hyperplasia in rats.