Learn More
Medchemexpress LLC Dapoxetine hydrochloride | 129938-20-1 | 100.0% | 341.87 | C21H24ClNO | 10 MG
SDP
Catalog No. 5000298869
Encompass_Preferred
Dapoxetine hydrochloride is the hydrochloride salt of dapoxetine, an orally active selective serotonin reuptake inhibitor (SSRI) used in research on premature ejaculation and serotonin transporter pharmacology. It potently inhibits serotonin uptake and has weaker activity at norepinephrine and dopamine transporters. Supplied as a white to off-white solid with high analytical purity and defined storage recommendations.
  • Serotonin transporter (SERT) inhibitor (5-HT uptake IC50 ≈ 1.12 nM)
  • Shows measurable norepinephrine and dopamine activity
  • High purity suitable for analytical and research use
  • White to off-white solid, stable when stored sealed away from moisture
  • Available in small mass quantities for laboratory studies
Catalog No. 50-002-98869 Supplier Medchemexpress LLC Supplier No. HYB0304A10MG
Dapoxetine hydrochloride is the hydrochloride salt of dapoxetine, an orally active selective serotonin reuptake inhibitor (SSRI) used in research on premature ejaculation and serotonin transporter pharmacology. It potently inhibits serotonin uptake and has weaker activity at norepinephrine and dopamine transporters. Supplied as a white to off-white solid with high analytical purity and defined storage recommendations.
  • Serotonin transporter (SERT) inhibitor (5-HT uptake IC50 ≈ 1.12 nM)
  • Shows measurable norepinephrine and dopamine activity
  • High purity suitable for analytical and research use
  • White to off-white solid, stable when stored sealed away from moisture
  • Available in small mass quantities for laboratory studies