Learn More
Medchemexpress LLC Dapoxetine hydrochloride | 129938-20-1 | 99.96% | 50 MG
SDP
Catalog No. 5000406666
Encompass_Preferred
Dapoxetine hydrochloride is an orally active and selective serotonin reuptake inhibitor (SSRI) used for premature ejaculation (PE) research. It binds to 5-HT, norepinephrine, and dopamine reuptake transporters.
  • Inhibits 5-HT, norepinephrine, and dopamine uptake
  • Can be used for premature ejaculation (PE) research
  • Oral gavage significantly inhibits testosterone-mediated increase in prostate weight in rats
Catalog No. 50-004-06666 Supplier Medchemexpress LLC Supplier No. HYB0304A50MG
Dapoxetine hydrochloride is an orally active and selective serotonin reuptake inhibitor (SSRI) used for premature ejaculation (PE) research. It binds to 5-HT, norepinephrine, and dopamine reuptake transporters.
  • Inhibits 5-HT, norepinephrine, and dopamine uptake
  • Can be used for premature ejaculation (PE) research
  • Oral gavage significantly inhibits testosterone-mediated increase in prostate weight in rats