Learn More
TARGETMOL CHEMICALS INC Dapoxetine hydrochloride 200MG
SDP
Catalog No. 502876353
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Dapoxetine hydrochloride (LY-210448 hydrochloride) is a selectivity short-acting serotonin reuptake inhibitor, used for the therapy of premature ejaculation. Purity 99%
Catalog No. 50-287-6353 Supplier TARGETMOL CHEMICALS INC Supplier No. T6461200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Dapoxetine hydrochloride (LY-210448 hydrochloride) is a selectivity short-acting serotonin reuptake inhibitor, used for the therapy of premature ejaculation. Purity 99%